ContactUsLinkComponent
Flow meters for KrosFlo® systems
Flow meters for KrosFlo® systems
Catalog # SPECACM3-FMH-01N
Flow meters for KrosFlo® systems
Catalog # SPECACM3-FMH-01N
Specifications
  • Flow rate:
    0,2 - 4,5 l/min
  • Description:
    Permeate flow meter
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Permeate flow meters for use with tangential flow KrosFlo® systems.