ContactUsLinkComponent
Absorbing sheet for medical packaging
Absorbing sheet for medical packaging
Catalog # DAKL460583
Supplier:  DAKLAPACK
Absorbing sheet for medical packaging
Catalog # DAKL460583
Supplier:  DAKLAPACK
Specifications
  • Description:
    Absorbing sheet, L×W 200×300 mm, absorbing capacity 150 ml
  • Pk:
    100
Specifications
About this item
These non-transparent absorbing materials are part of the packaging instruction P650. In emergencies the material will absorb all the liquid that has to stay inside the packaging. The absorbing sheets are on one side provided with a fluid-tight base. That ensures that the outside will stay clean and dry.