Order Entry
Switzerland
ContactUsLinkComponent
Water purification systems, UltraClear
Water purification systems, UltraClear
Certificates
Water purification systems, UltraClear

Specifications

  • Catalog No:
  • SGWA2002-D/80
  • 171-0042
  • 171-0167
  • 171-0166
  • 171-0044
  • Model:
  • 2002-D/80-SW
  • 2001-D-SW
  • 2002-D/60-SW
  • 2001-D/60-SW
  • 2002-D-SW
  • Feed water:
  • Tap water feed
  • Tap water feed
  • Tap water feed
  • Tap water feed
  • Tap water feed
  • Flow rate:
  • 1,8 l/min
  • 1,8 l/min
  • 1,8 l/min
  • 1,8 l/min
  • 1,8 l/min
  • Conductivity:
  • 0,055 μS/cm
  • 0,055 μS/cm
  • 0,055 μS/cm
  • 0,055 μS/cm
  • 0,055 μS/cm
  • TOC content:
  • <1 ppb
  • 5 - 10 ppb
  • <1 ppb
  • 5 - 10 ppb
  • <1 ppb
  • Resistivity at +25 °C:
  • 18,2 MΩ/cm at 25 °C
  • 18,2 MΩ/cm at 25 °C
  • 18,2 MΩ/cm at 25 °C
  • 18,2 MΩ/cm at 25 °C
  • 18,2 MΩ/cm at 25 °C
  • Bacteria:
  • <1 CFU/ml
  • <1 CFU/ml
  • <1 CFU/ml
  • <1 CFU/ml
  • <1 CFU/ml
  • Particles:
  • <1/ml at 0,1 µm
  • <1/ml at 0,1 µm
  • <1/ml at 0,1 µm
  • <1/ml at 0,1 µm
  • <1/ml at 0,1 µm
  • W×D×H:
  • 1340×340×580 mm
  • 560×320×530 mm
  • 530×900×320 mm
  • 900×320×530 mm
  • 530×560×320 mm
  • Weight:
  • 57 kg
  • 41 kg
  • 45 kg
  • 44 kg
  • 42 kg

Specifications

Cartridges

Product Information