Disposable particle filtering respirators
Fournisseur: HONEYWELL SAFETY
Certificats
À propos de cet article
Ultra-light, disposable FFP half masks with elastic straps. Ideal to protect against dusts, mists and fumes.
- Upper part pre-formed (no nose-bridge) for a quick and easy adjustment
- Great inner volume to avoid any oppression on face and to ease communication
- Ultra-soft inner veil to offer a soft and pleasant contact to the skin and improve the breathing comfort
- Twin supple and resistant elastics, stapled on ear tabs to avoid any risk of irritation and of leakage in the filtering area
- No latex, no silicone, no PVC
- Filtration Efficiency >99,9%
Ideal for use in textile, construction, iron and steel industries.
Certifications: EN 149:2001 + A1: 2009
Frequently Bought Together