ContactUsLinkComponent
Scrub pads, ErgoSCRUB®
Scrub pads, ErgoSCRUB®
Catalog # FOAMFT900
Supplier:  FOAMTEC
Scrub pads, ErgoSCRUB®
Catalog # FOAMFT900
Supplier:  FOAMTEC
Specifications
  • Description:
    ErgoSCRUB® pad, soft handle, disposable
  • Pk:
    10
Specifications
About this item
ErgoSCRUB® is a soft ergonomically correct scrubbing tool for easy scrubbing of implant tools, large vacuum chambers, ceramic domes, and other spherical surfaces.
  • Disposable
  • Easy to grip
  • Flexible enough to clean spherical surfaces, such as domes
  • Enables faster cleaning of large tool surfaces
Suitable for chamber cleaning, CVD and CMP applications and for wafer handling.