UltraMOP™III cleanroom wall mop heads
Supplier: FOAMTEC
Certificates
About this item
UltraMOP™ III mop heads are constructed of a snag-resistant, polyester fabric laminated to an absorbent polyurethane foam employing the patent pending Bump-PROOF® design. The Bump-PROOF® design improves cleanliness and durability as the durable fabric extends over the front and rear edges making it ideal for ISO class 4 and 5 clean rooms.
- Compatible with gamma, autoclave and EtO sterilisation methods
- Constructed of a proprietary, super, fine pore polyurethane foam that is ideal to laydown a precise volume of disinfectant cleaner
- Laminated fabric covers the leading and trailing edge preventing abrasion related particle contamination
- Product traceability: available with lot Certification of Conformance to ease audit and validation procedures
- Simple to use: bump-proof wrap-around design eliminates the need for protective polyester sock
- Robust cleaning: microfiber versions remove stubborn contamination and are excellent where dry floors are desired
Frequently Bought Together