Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Ludwig Schneider
Precision thermometers
Supplier:  Ludwig Schneider
Certificates
Precision thermometers
Precision thermometers
5 Options Available Below
Product Details & Documents
These precision laboratory thermometers are suitable for official certification.
  • Total immersion
  • White back or colourless prismatic capillary form
  • Filing with toluol or alcohol
  • Stabilisation of the thermometer zero point by molecular-constant, artificial ageing
Enclosed or stem type; the stem types are acid and base resistant. These thermometers can be used for application in science and research.
Conforms to DIN 12778.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Conform to DIN 12778.
Ordering information: Available with works certificate or with DAkkS calibration certificate.
Product Family Options
Without certificate
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1032302
    Enclosed type, with colourless, prismatic capillary form
    Enclosed type, with colourless, prismatic capillary form
    −100...+30 °C
    1,0 °C
    305 mm
    ALCH
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 49.00
    SFr. 49.00
    1 * 1 ST
       SFr. 49.00
  • SCNL1032303
    Enclosed type, with colourless, prismatic capillary form
    Enclosed type, with colourless, prismatic capillary form
    −50...+50 °C
    1,0 °C
    305 mm
    Toluol
    Total
    9 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 74.90
    SFr. 74.90
    1 * 1 ST
       SFr. 74.90
  • SCNL1032306
    Enclosed type, with colourless, prismatic capillary form
    Enclosed type, with colourless, prismatic capillary form
    0...+250 °C
    1,0 °C
    350 mm
    Mercury
    Total
    9 ±1 mm
    Discontinued
  • SCNL1032402
    Stem type, with white back, round capillary form
    Stem type, with white back, round capillary form
    −100...+30 °C
    1,0 °C
    305 mm
    ALCH
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 47.90
    SFr. 47.90
    1 * 1 ST
       SFr. 47.90
  • SCNL1032403
    Stem type, with white back, round capillary form
    Stem type, with white back, round capillary form
    −50...+50 °C
    1,0 °C
    305 mm
    Toloul
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 38.00
    SFr. 38.00
    1 * 1 ST
       SFr. 38.00
With certificate
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • SCNL1032402/04/3P
    Stem type with white back, round capillary form, calibrated by three points
    Stem type with white back, round capillary form, calibrated by three points
    −100...+30 °C
    1,0 °C
    305 mm
    ALCH
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 323.00
    SFr. 323.00
    1 * 1 ST
       SFr. 323.00