Topical irrigation solution, Aqua B. Braun
Supplier: B. Braun
Certificates
About this item
Sterile and pyrogen-free topical irrigation solution. Applications include moistening of tissue and cleansing of the operating area during surgical procedures, rinsing of wounds in the event of injuries and burns, cleansing during stoma care, moistening of wound tamponades, cloths, bandages, and dressings. Also used for cleansing of operating instruments and accessories or filling of humidifiers and vaporisers.
- No more troublesome, time consuming decanting of irrigation solutions and aspirating the solution into a syringe
- Precise irrigation from soft container with the spray nozzle
Frequently Bought Together