Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Grounders
Wrist Grounders
ESD wrist strap, accessory for ESD work stations
ESD wrist strap, accessory for ESD work stations
ESD wrist strap, accessory for ESD work stations
SOVE860513-00
:
TRESTON
ESD wrist strap, accessory for ESD work stations
SOVE860513-00
:
TRESTON
Colour:
Dark blue
Description:
ESD wrist band, fabric, with press-stud (7 mm)
Seq:
01
Pk:
1