Cryoboxes
: NEOLAB
Cryo resistant cardboard.
- Water repellent
- Llid has a marking area on one of the sides
For storage of tissue culture tubes of 15 and 50 ml capacities for cryogenic deep freezers.
Product Family Options
- DescriptionCompartmentsForW×D×H ext.
- without grid insert-Tissue culture tubes 15/50 ml122×122×128 mm1 * 1 STSFr. 14.20
- without grid insert-Tissue culture tubes 15/50 ml148×148×128 mm1 * 1 STSFr. 16.20