Water purification system, AFS® 10E/15E
Supplier: Merck
Certificates
About this item
Economical water system with Elix® technology, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 L/min. For clinical analysers with pure water needs up to 300 L daily.
- Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality
- Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
- No resin cylinders to replace: Lower running costs
- Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
- Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
Patented Elix® technology provides constant quality water without the need for softeners or conditioning cartridges, which means reduced maintenance― and less analyser downtime.
Specifications
- Catalog No:
- MILLZAFS10E0SZ
- MILLZAFS15E0SZ
- Model:
- AFE® 10E
- AFE® 15E
- Feed water:
- Portable tap water at a maximum pressure of 6 bar
- Portable tap water at a maximum pressure of 6 bar
- Production flow rate:
- 10 L/h @ 25 °C
- 15 L/h @ 25 °C
- TOC content:
- < 30 ppb
- < 30 ppb
- Resistivity at +25 °C:
- >15 MΏ.cm
- >15 MΏ.cm
- Bacteria:
- < 1 CFU/ml
- < 1 CFU/ml
- W×D×H:
- 268×426×585 mm
- 268×426×585 mm
- Weight:
- 28 kg
- 28 kg
Specifications
Frequently Bought Together