Container, EUROBOX
Supplier: ZARGES LEICHTBAU
New Product
Certificates
About this item
Aluminium, with lid.
- Lid with stainless steel hinges and holding straps
- Blue stacking corners ensure safe stacking
- Corner beads for additional dimensional stability
Eurobox has high resistance against corrosion due to stainless steel fittings. The peripheral foamed seal in the lid protects against dust and splash water.
Frequently Bought Together