Surface disinfectant, concentrate, Mikrobac® Forte
Supplier: Bode
Certificates
About this item
Mikrobac® forte is suitable for the daily disinfecting maintenance cleaning of water-resistant surfaces in areas where odourless disinfection is preferred.
- Contains benzyl-C12-18-alkyldimethylammonium chlorides 199 mg/g, N-(3-aminopropyl)-N-dodecylpropane-1, 3-diamine 50 mg/g
- Bactericidal, fungicidal, tuberculocidal, virucidal against enveloped virus and MNV, adeno, polyoma and rotavirus
- pH value of 0,5% solution: approx 8
- Suitable for industrial kitchens and food-processing areas
Caution: * Available in Germany only.
Frequently Bought Together