Flame photometers, 360
Anbieter: SHERWOOD SCIENTIFIC INSTRUMENT
Zertifikate
Über diesen Artikel
Single channel flame photometer for analysis of five elements (sodium, potassium, lithium, calcium, barium) designed with front loading, unobstructed sample work area and removable tray for easy cleaning.
- Automatic ignition and flame optimisation
- Safety features including fail safe flame detection and shut off with low air pressure indicator
- High sensitivity with fine manual control of air and stable flame design
- Indicator on the front panel indicate the filter selected and the flame status
- Useful display 'HOLD' button to ensure that readings are not lost before record
Delivery: Supplied with Na, K, Ca, Ba and Li standard solutions (1000 ppm), reinforced nylon hose, tubing clip, drain tube, atomiser, nebuliser tubing and cleaning wire, PSU multi plug adapter and operating instructions.
Technische Daten
- Catalog No:
- SHEW36000209
- SHEW36000009
- Model:
- 360 Clinical
- 360 Industrial
- Reproducibility:
- (Na) Better than 1,5% CV @ 140 mmol/l
(K) Better than 1,5% CV @ 5 mmol/l
(K) Better than 1,5% CV @ 80 mmol/l
(Li) Better than 2,5% CV @ 1,5 mmol/l - CV 1,0% for 20 aliquots sampled for 20 seconds using 10 ppm K solution
CV 1,5% for 20 consecutive samples using 10 ppm K solution
- Linearity:
- (Na) ±2 mmol/l (K/Li) Up to ±0,2 mmol/l - Serum
(Na) ±4 mmol/l (K) Up to ±2 mmol/l - Urine - (Na/K/Li) ±2% at 3 ppm (Ca) ±2% at 100 ppm (Ba) ±2% at 1000 ppm
- W×D×H:
- 420×350×480 mm
- 420×350×480 mm
- Weight:
- 8,6 kg
- 8,6 kg
Technische Daten
Frequently Bought Together