Tank tool
About this item
The tank tool is designed to efficiently clean chambers, tanks and pipes from above or below. The articulated head allows curved frame to hug the outer wall of the tank as it moves downward to remove residue or apply disinfectant. The unit has a degree of swivel to ensure good contact with the surface being cleaned.
- Stainless steel frame adapter with swivel action
- Curved frame maintains contact while cleaning tanks
- Available in exterior and interior (reverse) configurations
- Designed for cleanroom use
Frame adapters hold MZSM series of replaceable mop heads. Covers available in a variety of fabrics.
Attaches directly to SSU series handles. Attaches to STAH and STAEH series via SSA-2 Adapter.
4 Options Available Below
Product Details & Documents
The tank tool is designed to efficiently clean chambers, tanks and pipes from above or below. The articulated head allows curved frame to hug the outer wall of the tank as it moves downward to remove residue or apply disinfectant. The unit has a degree of swivel to ensure good contact with the surface being cleaned.
- Stainless steel frame adapter with swivel action
- Curved frame maintains contact while cleaning tanks
- Available in exterior and interior (reverse) configurations
- Designed for cleanroom use
Frame adapters hold MZSM series of replaceable mop heads. Covers available in a variety of fabrics.
Attaches directly to SSU series handles. Attaches to STAH and STAEH series via SSA-2 Adapter.
Product Family Options
Product Information
- AvailabilityPrice
- 129-1072Reverse tank tool adapter, electropolished stainless steelReverse tank tool adapter, electropolished stainless steelLog in for availability1 * 1 STSFr. 577.00
- 129-1073Tank tool adapter, electropolished stainless steel
- MICRQPSLTT-14PTankTool™ Exterior Curve, threaded handle connector
- MICRQPSLRTT-14BTankTool™ Interior Curve, vertical threaded handle connectorTankTool™ Interior Curve, vertical threaded handle connectorLog in for availability1 * 1 STSFr. 213.00
Microfibre cover
Product Information
- Catalog #DescriptionLengthAvailabilityPrice
- SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide360 mmLog in for availability1 * 1 STSFr. 36.00
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..