Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Triethanolamine 1000 mg/l in water standard for ion chromatography
VWR®, Triethanolamine 1000 mg/l in water standard for ion chromatography
  85001.180
 :  
undefined
VWR®, Triethanolamine 1000 mg/l in water standard for ion chromatography
  85001.180
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Triethanolamine standard

 

 

 : Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.