Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Oxalate 1000 mg/L in water standard for ion chromatography
VWR®, Oxalate 1000 mg/L in water standard for ion chromatography
Catalog # 84986.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Oxalate 1000 mg/L in water standard for ion chromatography
Catalog # 84986.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Oxalate standard solution
  • Contents:
    H₂C₂O₄ in H₂O

Specifications

About this item

Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.