VWR®, Monomethylamine 1000 mg/L in water standard for ion chromatography
Katalognr. 84980.180
Anbieter: VWR Chemicals
CAS-Nummer:
Technische Daten
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Monomethylamine standard
Technische Daten
Über diesen Artikel
Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.