ContactUsLinkComponent
Inverted microscopes, AE31E
Inverted microscopes, AE31E
Supplier:  MOTIC
Certificates
undefined
Inverted microscopes, AE31E
Supplier:  MOTIC
Specifications
  • Catalog No:
  • MOTIP31FLUOR
  • 630-2521
  • 630-2518
  • 630-2519
  • MOTIP31S
  • MOTIP31LED
  • 630-2971
  • MOTIP31L
  • Head:
  • Siedentopf, 45° inclined
  • Siedentopf, 45° inclined
  • Siedentopf, 45° inclined
  • Eyepieces:
  • WF 10×/22 mm with pointer adjustment and rubber eyecup
  • WF 10×/22 mm with pointer adjustment and rubber eyecup
  • WF 10×/22 mm with pointer adjustment and rubber eyecup
  • Nosepiece:
  • Quintuple, inclined
  • Quintuple, inclined
  • Quintuple, inclined
  • Objectives:
  • 4×, ph10×, phLWD 20×, ph LWD 40×
  • 4×, ph10×, phLWD 20×, ph LWD 40×
  • 4×, ph10×, phLWD 20×, ph LWD 40×
  • Stage:
  • Hard coated with metal and glass stage inserts
  • Condenser:
  • N.A 0,30 with extra-long working distance (72 mm) and diaphragm
  • N.A 0,30 with extra-long working distance (72 mm) and diaphragm
  • N.A 0,30 with extra-long working distance (72 mm) and diaphragm
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Illumination:
  • Halogen (Köhler)
  • Halogen (Köhler)
  • Halogen (Köhler)
Specifications
Camera Adapters
Product Information