Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Baths
Shaking Water Baths
Accessories for Aqua pro range shaking water baths
Accessories for Aqua pro range shaking water baths
Specifications
Description:
Heat exchange coil
Seq:
60
Environmentally Preferable:
Pk:
1
For:
Source of constant cooling, allows bath to be operated at or below ambient, down to 0 °C, fits OLS26
Specifications
About this item
Product Details
Product Details