Orders LinkContactUsLinkComponent
Threaded adapters, BOLA
Threaded adapters, BOLA
Supplier:   Bohlender
Threaded adapters, BOLA
Supplier:  Bohlender
Certificates
Threaded adapters, BOLA
Threaded adapters, BOLA
2 Options Available Below
Product Details & Documents
Adaptors, made of PTFE, white, allow the use of BOLA multiple distributors for bottles with female thread GL 45 also on bottles with GL 32, GL 40 and S 40 threads.
  • Chemical resistant
  • Temperature resistant from −200 to +200 °C
Product Family Options
Product Information
  • Availability
    Price
  • BOHLH978-30
    Transition from GL 32 to GL 45
    Transition from GL 32 to GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 68.90
    SFr. 68.90
    1 * 1 ST
       SFr. 68.90
  • BOHLH978-40
    Transition from GL40/S40 to GL 45
    Transition from GL40/S40 to GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.40
    SFr. 26.40
    1 * 1 ST
       SFr. 26.40