Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cleaning brush

Cleaning brush

Foam, 60 mm, Plastic, 250 mm
Catalog # ROTHY110.1
Marketsource
Supplier:   Roth Carl
Cleaning brush
Catalog # ROTHY110.1
Supplier:  Roth Carl
Marketsource
Cleaning brush

Foam, 60 mm, Plastic, 250 mm

Price
SFr. 20.35
Log in for availability
1 * 1 ST
Product Details & Documents

Brushes with non-scratching PE-foam and plastic handle. Specially designed for cleaning conical laboratory glass and plastic containers.

  • Does not absorb liquids
  • Round head for small vessels
Specifications
  • Bristle material:
    Foam
  • Brush head Ø:
    60 mm
  • Handle material:
    Plastic
  • Total length:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.