Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Staining tray

Staining box with PP cover, TPX, 125×125×50 mm
Catalog # SEMA4332
Supplier:   SEMADENI
Staining tray
Catalog # SEMA4332
Supplier:  SEMADENI
Staining tray

Staining box with PP cover, TPX, 125×125×50 mm

Price
SFr. 38.10
Log in for availability
1 * 1 ST
Product Details & Documents

Transparent base with a well fitting PP lid.

  • Usable up to 70 °C
  • Stackable with two handles
  • Good chemical resistance

Suitable for dying, bleaching and fixing of gels and membranes susceptible electrophoresis.

Specifications
  • Description:
    Staining box with PP cover
  • Material:
    TPX
  • W×D×H:
    125×125×50 mm