ContactUsLinkComponent
Cleanroom papers, CRB18
Cleanroom papers, CRB18
  CONNPB2C001WHITE
undefined
Cleanroom papers, CRB18
  CONNPB2C001WHITE
  • Colour:
    White
  • Description:
    Computer paper, continuous feed, 241,3×279 mm
  • Pk:
    1.700
CRB-18 cleanroom papers are saturated with syntheic polymers which seals individual fibers into the sheet to minimize particle generation. The papers can be used for a variety of uses in the cleanroom, primarily note taking and record keeping.
  • Heat resistant for use in laser printers and photocopy machines
  • Autoclavable