Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bi-metallic dial thermometer

Bi-metallic dial thermometer

0…120 °C, 100 mm, 100 mm
Catalog # AMART110410
Supplier:   Amarell
Bi-metallic dial thermometer
Catalog # AMART110410
Supplier:  Amarell
Bi-metallic dial thermometer

0…120 °C, 100 mm, 100 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Bimetal dial thermometers made of brass. The scale cover made of glass is used in heating systems and indutrial plants.

  • Axial stem
  • Thread: R ½"
  • Quick and reliable temperature control
Specifications
  • Ø:
    100 mm
  • Length:
    100 mm
  • Range:
    0…120 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.