ContactUsLinkComponent
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Catalog # SCUE121501
undefined
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Catalog # SCUE121501
Specifications
  • Description:
    Octenisan® antibacterial shampoo and body wash, 150 ml bottle
  • Seq:
    01
  • Pk:
    30
Specifications
About this item
Mild and gentle antimicrobial shampoo and body wash. Suitable for all skin types, also for all over body washing to combat MRSA. Particularly suitable for hospital use in intensive care and infectious diseases units.
  • Contains specially selected skin care ingredients, gentle surfactants and octenidine dihydrochloride
  • Skin-neutral pH
  • Free of colourant and perfume
Caution: Use biocides safely. Always read the label and product information before use.
*The product(s) on this page are outside the scope of the EU Directive 2019/904