Surface disinfectant, perform® classic concentrate OXY
Catalog # SCUE122320
Supplier: SCHÜLKE & MAYR GMBH
Specifications
- Description:perform® classic concentrate OXY, surface disinfectant powder,
- Environmentally Preferable:
- Packed:250 × 40 g sachet per case
- Pk:250
Specifications
About this item
Powdered concentrate based on active oxygen suitable for surface disinfection.
- Material compatible
- Convenient single-use sachet
- Colour-free
- Bactericidal, virucidal (BVDV/Vaccinia), fungicidal and sporicidal
Caution: Use biocides safely. Always read the label and product information before use.