Surface Disinfectants, perform® advanced alcohol EP
Supplier: SCHÜLKE & MAYR GMBH
Certificates
About this item
Ready to use, alcohol-based, surface disinfectants for use in cleanrooms.
- Fast, effective and residue free
- 0,2 μm microbial filtered
- Colour- and fragrance-free
- Bactericidal, virucidal (BVDV, Vaccinia), fungicidal
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together