Order Entry
Switzerland
ContactUsLinkComponent
Surface disinfectant, perform® sterile alcohol EP
Surface disinfectant, perform® sterile alcohol EP
Catalog # SCUE121802
undefined
Surface disinfectant, perform® sterile alcohol EP
Catalog # SCUE121802

Specifications

  • Description:
    perform® sterile alcohol EP, for cleanrooms
  • Packed:
    500 ml spray bottles, double bagged
  • Pk:
    10

Specifications

About this item

Sterile, ready to use alcohol-based (~60% w/w ethanol/IPA), surface disinfectant for use in cleanrooms.

  • Fast and effective
  • Gamma irradiated and microbial filtered (0,2 μm)
  • Colour, perfume and residue free
  • Bactericidal, virucidal (BVDV/Vaccinia), fungicidal
Caution: Use biocides safely. Always read the label and product information before use.