Order Entry
Switzerland
ContactUsLinkComponent
 
Racks for centrifuge tubes
  SEMA3912
 :  SEMADENI
Racks for centrifuge tubes
  SEMA3912
 :  SEMADENI

 

  • For tubes:
    50 ml
  • No. of Holes:
    25
  • Colour:
    Blue
  • Seq:
    02
  • W×D×H:
    135×250×50 mm
  • Pk:
    1

 

 

Simple and inexpensive racks for 15 and 50 ml centrifuge tubes. Compatible with all standard universal tubes, respectively.

  • PP, blue