Disinfectants, ClearKlens Tego® 2000
Supplier: Diversey
Certificates
About this item
Highly effective, active amphoteric, surface disinfectant with a broad spectrum of antimicrobial activity, developed for use in sterile environments.
- Ready to use, 0,2 µm filtered, sterile filled and double-bagged
- Validated HPLC and titration test methods available
- Compatible with ClearKlens Cleansinald and ClearKlens Bi-Spore as a rotational process
Certifications: Passes CEN 1276, CEN 1650 and CEN 13697
Caution: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contact your local Avantor office.
Articles may not be available in all countries. Please contact your local Avantor office.