Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cleanroom bench mats

Cleanroom bench mats

Non-slip mats, Grey, Cleanroom mats, 600×450 mm, 2,5 mm
Catalog # DYCECZ06/BM60045GR
Supplier:   DYCEM
Cleanroom bench mats
Catalog # DYCECZ06/BM60045GR
Supplier:  DYCEM
Cleanroom bench mats

Non-slip mats, Grey, Cleanroom mats, 600×450 mm, 2,5 mm

Price
SFr. 657.00
Log in for availability
1 * 1 ST
Product Details & Documents

This non-slip mat prevents the movement and loss of small components used in laboratories to stabilise fragile equipment. Approved for use in all classes of cleanrooms, including microelectronics, micro mechanics, semi-conductor, pharmaceutical, biotech and medical devices.

  • Improves accuracy, precision and assembly time
  • No outgassing, silicone or DOP
  • Non-particulating - helmke drum test
  • Protects sensitive products by safely draining static discharge

Colour: Grey

Delivery information: Supplied with earthing stud.

Specifications
  • Type:
    Cleanroom mats
  • Colour:
    Grey
  • Description:
    Non-slip mats
  • L×W:
    600×450 mm
  • Thickness:
    2,5 mm