Hand disinfectant gels and solutions, Phagorub SPS
PHAG65023
- Description:Hand disinfectant solution, Phagorub SPS, 475 ml pump bottle
- Seq:05
- Pk:20
Hydroalcoholic gels and solutions for the disinfection of the hands. Special formulation enriched with moisturisers and emollients, ideal for sensitive skin.
- Bactericidal, mycobactericidal, fungicide, virucidal
- Ethanol based
- Gel viscosity suitable for hygienic controlled friction: 3 ml for 15 seconds
- Fragrance-free dyes
: Use biocides safely. Always read the label and product information before use.