ContactUsLinkComponent
Spatulas, Trulla type
Spatulas, Trulla type
Catalog # NICK9470
Supplier:  NICKEL ELECTRO
undefined
Spatulas, Trulla type
Catalog # NICK9470
Supplier:  NICKEL ELECTRO
Specifications
  • Description:
    Trulla spatulas, pattern type
  • Length:
    175 mm
  • Pk:
    5
Specifications
About this item
Stainless steel, PTFE XYLAN® coated. These spatulas feature a PTFE XYLAN® coating which provides a non stick surface to which very few solid or liquid substances will permanently adhere.
  • Stainless steel base: Spatula can withstand extreme temperatures and chemicals
  • PTFE coating provides chemical inertness, purity, low permeability, heat resistance, toughness, and non stick qualities
  • Excellent non wetting properties make cleaning very easy
  • Spatula can undergo steam or chemical sterilisation
Ideal for biology, microbiology and pharmaceutical applications.