Order Entry
Switzerland
ContactUsLinkComponent
Centrifuge bottles, PP/PE, with screw cap
Centrifuge bottles, PP/PE, with screw cap
Catalog # 215-0224
Supplier:  SEMADENI
undefined
Centrifuge bottles, PP/PE, with screw cap
Catalog # 215-0224
Supplier:  SEMADENI

Specifications

  • Ø×L:
    69,5×163,5 mm
  • Capacity:
    500 ml
  • Pk:
    1

Specifications

About this item

PPCO, translucent with PE screw cap.

  • Autoclavable
  • Excellent chemical resistance
  • Withstands forces up to 27500×g for 250 ml, 13500×g for 500 ml and 7100×g for 1000 ml