Jars, with slip lid
Supplier: SEMADENI
Certificates
About this item
PS, clear
- Good chemical resistance
- Temperature resistance from −10 to 70 °C
Ideal for sampling and storage of solid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together