Order Entry
Switzerland
ContactUsLinkComponent
Jerricans, with outlet connection
Jerricans, with outlet connection
Catalog # 215-0233
Supplier:  SEMADENI
undefined
Jerricans, with outlet connection
Catalog # 215-0233
Supplier:  SEMADENI

Specifications

  • Capacity:
    10 l
  • Seq:
    01
  • W×D×H:
    265×130×400 mm
  • Pk:
    1

Specifications

About this item

HDPE, translucent.

  • Good chemical resistance
  • Good temperature resistance

Universal jerrican for industry, trade and laboratories.