ContactUsLinkComponent
Quick release clamp, for SLV® reaction vessels
Quick release clamp, for SLV® reaction vessels
Catalog # BIBB295-02
Supplier:  SciLabware
undefined
Quick release clamp, for SLV® reaction vessels
Catalog # BIBB295-02
Supplier:  SciLabware
Specifications
  • For:
    100 mm flanges, complete
  • Pk:
    1
Specifications
About this item
Made of stainless-steel.
  • For securing SVL® lids to reaction vessels
  • One piece construction
  • Can be secured and removed in a single movement with an adjustable locking tab