Trays, multipurpose, PP
Supplier: SEMADENI
Jump to:
About this item
Made of PP, white.
- Stable, durable, unbreakable
- Temperature resistant from −20 to +130 °C
- Smooth surfaces for easy cleaning
Multipurpose trays. Suitable for use as a tray for aggressive substances, sterilisation and photographic development.
6 Options Available Below
Product Details & Documents
Made of PP, white.
- Stable, durable, unbreakable
- Temperature resistant from −20 to +130 °C
- Smooth surfaces for easy cleaning
Multipurpose trays. Suitable for use as a tray for aggressive substances, sterilisation and photographic development.
Documents
SEMA3618_TDS_EN
Datasheet
Product Family Options
Product Information
- ColourMaterialW×D×HAvailabilityPrice