Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Containers, disposable, with cap

Containers, disposable, with cap

25 ml, 45 mm, 30 mm
Catalog # SEMA2988
Supplier:   SEMADENI
Containers, disposable, with cap
Catalog # SEMA2988
Supplier:  SEMADENI
Containers, disposable, with cap
Containers, disposable, with cap

25 ml, 45 mm, 30 mm

Price
SFr. 42.20
Log in for availability
1 * 100 ST
Product Details & Documents

Transparent cup made of PS with well fitting PE cap, universally applicable.

  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Ext.Ø:
    45 mm
  • Capacity:
    25 ml
  • Height:
    30 mm