Orders LinkContactUsLinkComponent
Guinea pig serum
Guinea pig serum
 :   Biowest
Guinea pig serum
 :  Biowest
Guinea pig serum
Guinea pig serum
Guinea pig serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Guinea pig serum is used as a blocking agent in immunoassays and controls.
Product Family Options
  • Description
    Sterility
    Serum format
  • Guinea pig serum
    Sterile filtered
    Frozen
    1 * 50 mL
    1 * 50 mL - SFr. 214.00
    SFr. 214.00
    1 * 50 mL
       SFr. 214.00
  • Guinea pig serum
    Sterile filtered
    Frozen
    1 * 100 mL
    1 * 100 mL - SFr. 390.00
    SFr. 390.00
    1 * 100 mL
       SFr. 390.00
  • Guinea pig serum
    Sterile filtered
    Frozen
    1 * 500 mL
    1 * 500 mL - SFr. 1’450.00
    SFr. 1’450.00
    1 * 500 mL
       SFr. 1’450.00