Orders LinkContactUsLinkComponent
Mouse serum
Mouse serum
Supplier:   Biowest
Mouse serum
Supplier:  Biowest
Certificates
Mouse serum
5 Options Available Below
Product Details & Documents
Mouse serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Mouse serum is used as a blocking agent in immunoassays and controls.
Documents
Product Family Options
Product Information
  • Sterility
    Serum format
    Availability
    Price
  • S2160-010
    Mouse serum
    Mouse serum
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - SFr. 52.60
    SFr. 52.60
    1 * 10 mL
       SFr. 52.60
  • S2160-020
    Mouse serum
    Mouse serum
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 20 mL
    1 * 20 mL - SFr. 92.90
    SFr. 92.90
    1 * 20 mL
       SFr. 92.90
  • S2160-050
    Mouse serum
    Mouse serum
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 172.00
    SFr. 172.00
    1 * 50 mL
       SFr. 172.00
  • Mouse serum
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 306.00
    SFr. 306.00
    1 * 100 mL
       SFr. 306.00
  • S2160-500
    Mouse serum
    Mouse serum
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 1’230.00
    SFr. 1’230.00
    1 * 500 mL
       SFr. 1’230.00