Orders LinkContactUsLinkComponent
Human plasma
Human plasma
Supplier:   Biowest
Human plasma
Supplier:  Biowest
Certificates
Human plasma
2 Options Available Below
Product Details & Documents
Human plasma is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Caution: Products of human origin should be considered potentially infectious and handled accordingly.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • S4180-500
    Human plasma, pooled
    Human plasma, pooled
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 452.00
    SFr. 452.00
    1 * 500 mL
       SFr. 452.00
  • S4180-100
    Human plasma, pooled
    Human plasma, pooled
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 157.00
    SFr. 157.00
    1 * 100 mL
       SFr. 157.00