ContactUsLinkComponent
Metallurgical microscopes, AE2000MET
Metallurgical microscopes, AE2000MET
Supplier:  MOTIC
Certificates
undefined
Metallurgical microscopes, AE2000MET
Supplier:  MOTIC
Specifications
  • Catalog No:
  • 630-2561
  • 630-2554
  • 630-2566
  • 630-2548
  • 630-2549
  • 630-2558
  • 630-2595
  • 630-2560
  • 630-2559
  • 630-2596
  • 630-2594
  • MOTI1101001704421
  • 630-2555
  • 630-2556
  • 630-2547
  • 630-2557
  • Head:
  • Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
  • Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
  • Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
  • Eyepieces:
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • WF 10×/20 mm with pointer adjustment
  • Nosepiece:
  • Quintuple, inclined
  • Quintuple, inclined
  • Quintuple, inclined
  • Stage:
  • 280×192 mm (movement: 50×50 mm)
  • 280×192 mm (movement: 50×50 mm)
  • 280×192 mm (movement: 50×50 mm)
  • Focusing system:
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Coaxial coarse and fine knobs
  • Illumination:
  • Halogen
  • Halogen
  • Halogen
Specifications
Related Information
Objectives
Product Information
Frequently Bought Together