ContactUsLinkComponent
Visual comparator disc, phosphate
Visual comparator disc, phosphate
Catalog # HACH9262100
Supplier:  Hach
CAS Number:  
undefined
Visual comparator disc, phosphate
Catalog # HACH9262100
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Phosphate, colour disc
  • Range:
    0 - 40 mg/l PO₄
Specifications
About this item
  • Fast acccurate comparison