Cleanroom wipes, pre-saturated, sterile, Sterile TechniSat®
Supplier: Texwipe
Certificates
About this item
These wipes constructed from a hydroentangled polyester/cellulose blend are pre-wetted with 70% isopropyl alcohol and 30% DIW (filtered through a 0,2 μm filter). Each package is gamma irradiated to 10⁻⁶ Sterility Assurance Level.
- Binder-free, hydroentangled polyester/cellulose substrate
- Packaging permits alcohol wipedown of exterior bag before introduction into sterile suites
- Offers low levels of solvent extractables and particle generation
- Ideal for wiping gloved hands
Ideal for alcohol wiping in sterile environments in pharmaceutical aseptic fill areas, biotechnology manufacturing facilities, microbiological laboratories, sterile suites and prep rooms.
Packaging: Packaged in a resealable FlexPack.
Frequently Bought Together