Stirrer shafts, double-moon shape, BOLA
Supplier: Bohlender
Certificates
About this item
Stirrer shafts made of PTFE-jacketed stainless steel, two each tilting half-moon stirrer blades with double-sided groove and access for the stirrer shaft completely made of PTFE.
- Chemical resistant
- Temperature resistance from –200 to +250 °C
- Creates tangential flow with little turbulence
- With chucking diameter of 8 mm
Half-moon blade is ideal for stirring in round-bottom flasks with ground joint neck.
Frequently Bought Together