Orders LinkContactUsLinkComponent
Ball valves
Ball valves
Ball valves, with an inner and outer thread, PP, 3/4" NW 20
Catalog # 229-7490
Supplier:   SEMADENI
Ball valves
Catalog # 229-7490
Supplier:  SEMADENI
Ball valves
Ball valves
Ball valves, with an inner and outer thread, PP, 3/4" NW 20
Price
SFr. 117.00
Log in for availability
1 * 1 ST
Product Details & Documents
Housing and ball made of PP, ball seal made of PTFE. These ball valves has a easy running, strong industrial design.
  • Operating pressure of 6 kg/cm² at max. 80 °C
  • Excellent chemical resistance
Covered both sides with gas thread for direct installation or via connection for hose.
Specifications
  • Description:
    Ball valves, with an inner and outer thread, PP, 3/4" NW 20