Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

 :   SEMADENI
Storage Bins
 :  SEMADENI
 
Storage Bins
Storage Bins

 

 

Made of PP.

  • Secure stacking
  • Made of high quality material and are dimensionally stable
  • Supporting edge and smooth inner surfaces for easy cleaning

Storage bins with large labelling field for content description.

Product Family Options

 

  • Colour
    W×D×H
    Material
     
     
  • Red
    165×105×75 mm
    PP
       
       
    1 * 1 ST
    1 * 1 ST - SFr. 3.20
    SFr. 3.20
    1 * 1 ST
       SFr. 3.20

  • Blue
    165×105×75 mm
    PP
       
       
    1 * 1 ST
    1 * 1 ST - SFr. 4.00
    SFr. 4.00
    1 * 1 ST
       SFr. 4.00

  • Blue
    230×150×125 mm
    PP
       
       
    1 * 1 ST
    1 * 1 ST - SFr. 6.60
    SFr. 6.60
    1 * 1 ST
       SFr. 6.60

  • Blue
    345×213×200 mm
    PP
       
       
    1 * 1 ST
    1 * 1 ST - SFr. 14.60
    SFr. 14.60
    1 * 1 ST
       SFr. 14.60

  • Blue
    500×315×200 mm
    PP
       
       
    1 * 1 ST
    1 * 1 ST - SFr. 31.00
    SFr. 31.00
    1 * 1 ST
       SFr. 31.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.