Orders LinkContactUsLinkComponent
Hydrometers
Hydrometers
Supplier:   Amarell
Hydrometers
Supplier:  Amarell
Certificates
Hydrometers
10 Options Available Below
Product Details & Documents
Density hydrometers with reference temperature of 15 °C (without thermometer).
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • AMARH827130
    Hydrometer
    Hydrometer
    600 - 650 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827132
    Hydrometer
    Hydrometer
    650 - 700 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827134
    Hydrometer
    Hydrometer
    700 - 750 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827136
    Hydrometer
    Hydrometer
    750 - 800 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827138
    Hydrometer
    Hydrometer
    800 - 850 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827140
    Hydrometer
    Hydrometer
    850 - 900 Kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827142
    Hydrometer
    Hydrometer
    900 - 950 Kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827144
    Hydrometer
    Hydrometer
    950 - 1000 Kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57.90 SFr.
    57.90 SFr.
    1 * 1 ST
       57.90 SFr.
  • AMARH827146
    Hydrometer
    Hydrometer
    1000 - 1050 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59.60 SFr.
    59.60 SFr.
    1 * 1 ST
       59.60 SFr.
  • AMARH827148
    Hydrometer
    Hydrometer
    1050 - 1100 kg/m³
    0,5 kg/m³
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59.60 SFr.
    59.60 SFr.
    1 * 1 ST
       59.60 SFr.